Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CNOT7 SIRNA Assays ›
Gene Symbol:
CNOT7
Gene Name:
CCR4-NOT transcription complex subunit 7
Gene Aliases:
CAF-1, CAF1, Caf1a, hCAF-1
Chromosome Location:
Chr.8: 17226030 - 17246909 on Build GRCh38
UniGene ID:
Hs.587465
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
3439
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CNOT7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001322087.1
NP_001309016.1
5
743
NM_001322088.1
NP_001309017.1
4
528
NM_001322089.1
NP_001309018.1
4
734
NM_001322090.1
NP_001309019.1
5
743
NM_001322091.1
NP_001309020.1
4
528
NM_001322092.1
NP_001309021.1
5
603
NM_001322093.1
NP_001309022.1
5
734
NM_001322094.1
NP_001309023.1
4
668
NM_001322095.1
NP_001309024.1
5
772
NM_001322096.1
NP_001309025.1
5
849
NM_001322097.1
NP_001309026.1
5
709
NM_001322098.1
NP_001309027.1
5
772
NM_001322099.1
NP_001309028.1
5
849
NM_001322100.1
NP_001309029.1
6
707
NM_013354.6
NP_037486.2
4
668
NM_054026.3
NP_473367.2
4
668
XM_005273481.2
XP_005273538.1
5
774
GenBank mRNA
AF086915.1
-
4
601
AK001209.1
-
4
584
AK021808.1
-
4
611
AK023466.1
-
4
637
AK225893.1
-
4
616
AL528251.3
-
4
625
AL833642.1
-
4
507
BC007315.2
-
4
599
2989585
BC060852.1
-
4
628
5273703
BC070187.1
-
4
614
4738218
BI458088.1
-
5
599
BI552176.1
-
4
534
BT006685.1
-
3
338
L46722.1
-
4
520
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001271543, NM_011135, NM_001271542, AK166449, AK147048, and others.
Panther Classification:
Molecular Function -
mRNA polyadenylation factor
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA poly(A) tail shortening
deadenylation-dependent decapping of nuclear-transcribed mRNA
carbohydrate metabolic process
transcription, DNA-templated
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
signal transduction
positive regulation of cell proliferation
negative regulation of cell proliferation
negative regulation of gene expression
gene silencing by RNA
cytoplasmic mRNA processing body assembly
gene silencing by miRNA
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
positive regulation of transcription from RNA polymerase II promoter
positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
positive regulation of mRNA catabolic process
RNA phosphodiester bond hydrolysis, exonucleolytic
positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
3'-5'-exoribonuclease activity
transcription factor activity, sequence-specific DNA binding
RNA binding
exoribonuclease activity
poly(A)-specific ribonuclease activity
signal transducer activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline