Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290325.1 | NP_001277254.1 | 1 | 839 | ||
NM_001290326.1 | NP_001277255.1 | 1 | 839 | |||
NM_032557.6 | NP_115946.2 | 1 | 839 | |||
XM_011532360.2 | XP_011530662.1 | 1 | 327 | |||
GenBank mRNA | AK093754.1 | - | 1 | 62 | ||
AK126943.1 | - | 2 | 667 | |||
AL833976.1 | - | 1 | 811 | |||
BC039115.1 | - | 1 | 839 | 5258804 | ||
BC068975.1 | - | 1 | 786 | 6067501 |