Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032559.4 | NP_115948.4 | 1 | 857 | ||
GenBank mRNA | AF333335.2 | - | 1 | 835 | ||
AK057272.1 | - | 1 | 834 | |||
AK308265.1 | - | 2 | 436 | |||
BC033802.1 | - | 1 | 795 | 5170969 |