Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCE3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032563.1 | NP_115952.1 | 2 | 504 | ||
GenBank mRNA | AB048288.1 | - | 2 | 504 |