Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256267.1 | NP_001243196.1 | 3 | 597 | ||
NM_001256268.1 | NP_001243197.1 | 3 | 766 | |||
NM_032578.3 | NP_115967.2 | 2 | 702 | |||
NR_045663.3 | 2 | 702 | ||||
XM_017016833.1 | XP_016872322.1 | 3 | 1038 | |||
XM_017016834.1 | XP_016872323.1 | 2 | 496 | |||
GenBank mRNA | AF328296.1 | - | 2 | 897 | ||
AK027343.1 | - | 2 | 640 | |||
AL834247.2 | - | 2 | 624 | |||
BC142609.1 | - | 2 | 574 | |||
BC144334.1 | - | 2 | 464 | |||
BX647637.1 | - | 2 | 634 | |||
BX647640.1 | - | 3 | 591 | |||
BX648564.1 | - | 4 | 1030 |