Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172674.1 | NP_001166145.1 | 4 | 630 | ||
NM_001172675.1 | NP_001166146.1 | 4 | 310 | |||
NM_032584.2 | NP_115973.2 | 4 | 307 | |||
XM_005259335.4 | XP_005259392.1 | 4 | 629 | |||
XM_005259336.4 | XP_005259393.1 | 4 | 419 | |||
XM_017027384.1 | XP_016882873.1 | 4 | 649 | |||
XM_017027385.1 | XP_016882874.1 | 4 | 333 | |||
XM_017027386.1 | XP_016882875.1 | 3 | 255 | |||
GenBank mRNA | AK096623.1 | - | 4 | 632 | ||
AK289994.1 | - | 4 | 294 | |||
AK314603.1 | - | 4 | 291 | |||
AY029765.1 | - | 4 | 269 | |||
BC136255.1 | - | 3 | 259 | 9020242 | ||
BC143412.1 | - | 3 | 262 |