Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261828.1 | NP_001248757.1 | 3 | 853 | ||
NM_032597.4 | NP_115986.3 | 3 | 853 | |||
NR_049732.1 | 3 | 853 | ||||
NR_049734.1 | 3 | 853 | ||||
GenBank mRNA | AF367473.2 | - | 3 | 364 | ||
AK314674.1 | - | 3 | 563 | |||
AY094610.1 | - | 3 | 437 | |||
BC107431.1 | - | 3 | 358 | |||
DB450359.2 | - | 3 | 361 | |||
DB727789.1 | - | 3 | 371 |