Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170695.1 | NP_001164166.1 | 6 | 752 | ||
NM_001170696.1 | NP_001164167.1 | 5 | 623 | |||
NM_001170697.1 | NP_001164168.1 | 6 | 758 | |||
NM_001170698.1 | NP_001164169.1 | 6 | 773 | |||
NM_001170699.1 | NP_001164170.1 | 6 | 992 | |||
NM_001321336.1 | NP_001308265.1 | 6 | 1042 | |||
NM_001321337.1 | NP_001308266.1 | 6 | 1090 | |||
NM_032598.4 | NP_115987.2 | 6 | 814 | |||
GenBank mRNA | AF367472.1 | - | 6 | 706 | ||
AK295299.1 | - | 6 | 1042 | |||
AK301892.1 | - | 6 | 958 | |||
AY032684.1 | - | 5 | 572 | |||
AY035867.1 | - | 6 | 755 | |||
AY035868.1 | - | 6 | 814 | |||
BC029483.1 | - | 6 | 699 | 5271286 |