Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004434.2 | NP_001004434.1 | 8 | 1327 | ||
NM_032513.4 | NP_115902.1 | 7 | 1180 | |||
GenBank mRNA | AF370409.1 | - | 8 | 1174 | ||
AK023491.1 | - | 3 | 349 | |||
AK023504.1 | - | 3 | 269 | |||
BC006251.2 | - | 7 | 1155 | 3948165 |