Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIRREL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301097.1 | NP_001288026.1 | 14 | 2038 | ||
NM_032531.3 | NP_115920.1 | 15 | 2074 | |||
XM_011543026.2 | XP_011541328.1 | 15 | 1893 | |||
XM_011543027.2 | XP_011541329.1 | 16 | 2354 | |||
XM_011543028.2 | XP_011541330.1 | 16 | 2213 | |||
XM_011543030.2 | XP_011541332.1 | 15 | 1815 | |||
XM_011543031.2 | XP_011541333.1 | 16 | 2273 | |||
XM_011543032.2 | XP_011541334.1 | 14 | 1779 | |||
XM_011543033.1 | XP_011541335.1 | 15 | 2243 | |||
XM_017018418.1 | XP_016873907.1 | 15 | 2444 | |||
XM_017018419.1 | XP_016873908.1 | 15 | 2318 | |||
XM_017018420.1 | XP_016873909.1 | 16 | 2138 | |||
GenBank mRNA | AB058770.1 | - | 15 | 1963 | ||
AF480410.1 | - | 15 | 1713 | |||
AK300108.1 | - | 5 | 457 | |||
AY358760.1 | - | 4 | 250 | |||
BC101775.1 | - | 15 | 1799 | 8069281 | ||
BC101801.1 | - | 15 | 1799 | 8069307 |