Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290325.1 | NP_001277254.1 | 7 | 1951 | ||
NM_001290326.1 | NP_001277255.1 | 8 | 2067 | |||
NM_032557.6 | NP_115946.2 | 7 | 1951 | |||
XM_011532360.2 | XP_011530662.1 | 6 | 1337 | |||
GenBank mRNA | AB067478.1 | - | 6 | 632 | ||
AK057992.1 | - | 4 | 475 | |||
AK093754.1 | - | 8 | 1290 | |||
AK126943.1 | - | 8 | 1779 | |||
AL833976.1 | - | 7 | 1923 | |||
BC039115.1 | - | 7 | 1951 | 5258804 | ||
BC068975.1 | - | 7 | 1898 | 6067501 |