Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318124.1 | NP_001305053.1 | 4 | 715 | ||
NM_001318125.1 | NP_001305054.1 | 3 | 629 | |||
NM_032562.4 | NP_115951.2 | 4 | 718 | |||
XM_005270244.4 | XP_005270301.1 | 4 | 931 | |||
GenBank mRNA | AF339053.1 | - | 4 | 677 | ||
AF349540.1 | - | 4 | 618 | |||
AK311349.1 | - | 3 | 616 | |||
AY358032.1 | - | 4 | 651 | |||
BC093996.1 | - | 4 | 640 | 7939841 | ||
BC111946.1 | - | 4 | 640 | 8327414 | ||
BC143532.1 | - | 4 | 637 | 9052041 |