Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000545.6 | NP_000536.5 | 3 | 868 | ||
NM_001306179.1 | NP_001293108.1 | 3 | 868 | |||
XM_005253931.3 | XP_005253988.1 | 3 | 921 | |||
GenBank mRNA | AK298422.1 | - | 3 | 742 | ||
BC104908.1 | - | 3 | 687 | 8143911 | ||
BC104910.1 | - | 3 | 687 | 8143913 | ||
BC143483.1 | - | 3 | 687 | |||
HM116541.1 | - | 3 | 810 | |||
HM116542.1 | - | 3 | 810 | |||
HM116547.1 | - | 3 | 810 | |||
HM116548.1 | - | 3 | 810 | |||
HM116552.1 | - | 4 | 427 | |||
HM116558.1 | - | 3 | 785 | |||
HM449088.1 | - | 3 | 690 | |||
HM449089.1 | - | 2 | 490 | |||
M57732.1 | - | 3 | 690 | |||
X71346.1 | - | 3 | 863 | |||
X71347.1 | - | 3 | 798 |