Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172674.1 | NP_001166145.1 | 5 | 2250 | ||
NM_001172675.1 | NP_001166146.1 | 5 | 1930 | |||
NM_032584.2 | NP_115973.2 | 5 | 1927 | |||
XM_005259335.4 | XP_005259392.1 | 5 | 2249 | |||
XM_005259336.4 | XP_005259393.1 | 5 | 2039 | |||
XM_017027384.1 | XP_016882873.1 | 5 | 2269 | |||
XM_017027385.1 | XP_016882874.1 | 5 | 1953 | |||
XM_017027386.1 | XP_016882875.1 | 4 | 1875 | |||
GenBank mRNA | AK095593.1 | - | 1 | 1488 | ||
AK096623.1 | - | 5 | 2252 | |||
AK289994.1 | - | 5 | 1914 | |||
AK314603.1 | - | 5 | 1911 | |||
AL713691.1 | - | 1 | 909 | |||
AY029765.1 | - | 5 | 1889 | |||
BC136255.1 | - | 4 | 1879 | 9020242 | ||
BC143412.1 | - | 4 | 1882 |