Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032588.3 | NP_115977.2 | 8 | 1138 | ||
XM_017002559.1 | XP_016858048.1 | 8 | 1138 | |||
GenBank mRNA | AF353673.1 | - | 8 | 1117 | ||
AJ276484.1 | - | 8 | 1121 | |||
AK056942.1 | - | 8 | 1134 | |||
AK297820.1 | - | 8 | 1050 | |||
BC080529.1 | - | 8 | 1116 | 4862321 |