Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170695.1 | NP_001164166.1 | 6, 7 | 856 | ||
NM_001170696.1 | NP_001164167.1 | 5, 6 | 727 | |||
NM_001170697.1 | NP_001164168.1 | 6, 7 | 862 | |||
NM_001170698.1 | NP_001164169.1 | 6, 7 | 877 | |||
NM_001170699.1 | NP_001164170.1 | 6, 7 | 1096 | |||
NM_001321336.1 | NP_001308265.1 | 6, 7 | 1146 | |||
NM_001321337.1 | NP_001308266.1 | 6, 7 | 1194 | |||
NM_032598.4 | NP_115987.2 | 6, 7 | 918 | |||
GenBank mRNA | AF367472.1 | - | 6, 7 | 810 | ||
AK295299.1 | - | 6, 7 | 1146 | |||
AK301892.1 | - | 6, 7 | 1062 | |||
AY032684.1 | - | 5, 6 | 676 | |||
AY035867.1 | - | 6, 7 | 859 | |||
AY035868.1 | - | 6, 7 | 918 | |||
BC029483.1 | - | 6, 7 | 803 | 5271286 |