Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCEE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032601.3 | NP_115990.3 | 2, 3 | 431 | ||
GenBank mRNA | AF364547.1 | - | 2, 3 | 404 | ||
BC020825.1 | - | 2, 3 | 396 | 4734422 | ||
BM271757.1 | - | 2, 3 | 453 |