Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129888.1 | NP_001123360.1 | 3 | 1125 | ||
NM_032687.3 | NP_116076.1 | 3 | 1161 | |||
XM_006716560.3 | XP_006716623.1 | 3 | 1378 | |||
XM_006716561.3 | XP_006716624.1 | 3 | 1330 | |||
GenBank mRNA | BC005073.1 | - | 3 | 1161 | 3533499 | |
BC056412.1 | - | 2 | 1439 | |||
BC065484.1 | - | 3 | 1125 | 4638402 |