Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032701.3 | NP_116090.2 | 7, 8 | 963 | ||
XM_005259338.3 | XP_005259395.1 | 6, 7 | 1288 | |||
XM_006723442.3 | XP_006723505.1 | 5, 6 | 696 | |||
XM_011527415.2 | XP_011525717.1 | 7, 8 | 1483 | |||
XM_017027388.1 | XP_016882877.1 | 5, 6 | 1744 | |||
GenBank mRNA | BC005842.2 | - | 5, 6 | 697 | 2821053 | |
BC019313.2 | - | 5, 6 | 692 | 2821053 | ||
BC044889.1 | - | 7, 8 | 920 | 3461265 |