Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF496 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032752.1 | NP_116141.1 | 9 | 2024 | ||
XM_005273328.3 | XP_005273385.1 | 11 | 3623 | |||
XM_005273330.3 | XP_005273387.1 | 10 | 3515 | |||
XM_011544310.1 | XP_011542612.1 | 12 | 3716 | |||
XM_011544312.1 | XP_011542614.1 | 11 | 3608 | |||
XM_017002586.1 | XP_016858075.1 | 13 | 3809 | |||
XM_017002587.1 | XP_016858076.1 | 13 | 3762 | |||
XM_017002588.1 | XP_016858077.1 | 12 | 3716 | |||
XM_017002589.1 | XP_016858078.1 | 12 | 3701 | |||
XM_017002590.1 | XP_016858079.1 | 11 | 3608 | |||
XM_017002591.1 | XP_016858080.1 | 10 | 3468 | |||
XM_017002594.1 | XP_016858083.1 | 6 | 1056 | |||
GenBank mRNA | AK024643.1 | - | 1 | 513 | ||
BC007263.1 | - | 9 | 2024 | 3051320 | ||
BC025794.1 | - | 8 | 1334 | |||
BX647480.1 | - | 2 | 5959 |