Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039970.1 | NP_001035059.1 | 4 | 589 | ||
NM_001278236.1 | NP_001265165.1 | 4 | 1423 | |||
NM_001278238.1 | NP_001265167.1 | 3 | 456 | |||
NM_001278239.1 | NP_001265168.1 | 3 | 456 | |||
NM_006906.1 | NP_008837.1 | 4 | 589 | |||
NM_032781.3 | NP_116170.3 | 4 | 544 | |||
XM_011520411.2 | XP_011518713.1 | 4 | 528 | |||
XM_017018434.1 | XP_016873923.1 | 4 | 528 | |||
XM_017018435.1 | XP_016873924.1 | 4 | 498 | |||
XM_017018436.1 | XP_016873925.1 | 3 | 434 | |||
XM_017018437.1 | XP_016873926.1 | 4 | 548 | |||
XM_017018438.1 | XP_016873927.1 | 2 | 213 | |||
XM_017018439.1 | XP_016873928.1 | 3 | 435 | |||
XM_017018440.1 | XP_016873929.1 | 4 | 528 | |||
XM_017018441.1 | XP_016873930.1 | 4 | 528 | |||
GenBank mRNA | AK090923.1 | - | 4 | 453 | ||
AK127312.1 | - | 4 | 1423 | |||
AK294673.1 | - | 3 | 253 | |||
AK295604.1 | - | 3 | 317 | |||
AK316270.1 | - | 3 | 241 | |||
AL832541.1 | - | 4 | 589 | |||
BC039897.1 | - | 3 | 252 | |||
BC064807.1 | - | 4 | 353 | 5726473 |