Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282736.1 | NP_001269665.1 | 2 | 798 | ||
NM_032812.8 | NP_116201.7 | 2 | 798 | |||
XM_011519750.2 | XP_011518052.1 | 2 | 1026 | |||
GenBank mRNA | AF378757.1 | - | 2 | 170 | ||
AK304591.1 | - | 3 | 682 | |||
AY358486.1 | - | 2 | 798 | |||
BC012885.1 | - | 2 | 404 | 3865073 |