Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFATC2IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032815.3 | NP_116204.3 | 6 | 931 | ||
GenBank mRNA | AF458593.1 | - | 6 | 941 | ||
AK027545.1 | - | 3 | 346 | |||
AK074761.1 | - | 6 | 921 | |||
AK297333.1 | - | 1 | 276 | |||
AK304606.1 | - | 1 | 198 | |||
AK316535.1 | - | 1 | 337 | |||
BC018311.1 | - | 4 | 435 | |||
BC021551.1 | - | 3 | 314 | |||
BC068007.1 | - | 3 | 171 | |||
BC080628.1 | - | 6 | 598 | |||
BC101741.1 | - | 6 | 991 | 8069247 | ||
BC112182.1 | - | 6 | 991 | 8327650 |