Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM220A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037163.1 | NP_001032240.1 | 2 | 653 | ||
GenBank mRNA | AF533707.1 | - | 2 | 648 | ||
AK093156.1 | - | 2 | 284 | |||
BC006110.1 | - | 2 | 653 | 3139887 | ||
BC140712.1 | - | 2 | 297 | 9021580 | ||
BC142977.1 | - | 2 | 669 | 8860408 |