Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYROXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032709.2 | NP_116098.2 | 3, 4 | 322 | ||
XM_011540293.2 | XP_011538595.1 | 4, 5 | 1744 | |||
XM_017016835.1 | XP_016872324.1 | 3, 4 | 283 | |||
XM_017016836.1 | XP_016872325.1 | 3, 4 | 283 | |||
XM_017016837.1 | XP_016872326.1 | 3, 4 | 283 | |||
XM_017016838.1 | XP_016872327.1 | 2, 3 | 1626 | |||
GenBank mRNA | AK074429.1 | - | 3, 4 | 283 | ||
AK075265.1 | - | 3, 4 | 260 | |||
AK225637.1 | - | 3, 4 | 276 | |||
AK295699.1 | - | 3, 4 | 295 | |||
BC006131.1 | - | 3, 4 | 285 | 3626506 | ||
DC361966.1 | - | 3, 4 | 323 |