Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282615.1 | NP_001269544.1 | 3 | 584 | ||
NM_001282616.1 | NP_001269545.1 | 4 | 679 | |||
NR_104220.1 | 2 | 346 | ||||
XM_017018865.1 | XP_016874354.1 | 4 | 634 | |||
GenBank mRNA | BC006440.1 | - | 3 | 501 |