Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032773.3 | NP_116162.1 | 4 | 649 | ||
XM_005269362.2 | XP_005269419.1 | 4 | 1020 | |||
XM_005269363.2 | XP_005269420.1 | 4 | 1020 | |||
XM_005269365.2 | XP_005269422.1 | 4 | 1020 | |||
XM_005269367.2 | XP_005269424.1 | 4 | 1020 | |||
XM_006713791.3 | XP_006713854.1 | 4 | 1020 | |||
XM_006713792.3 | XP_006713855.1 | 4 | 1020 | |||
XM_006713793.2 | XP_006713856.1 | 4 | 1020 | |||
XM_011513242.2 | XP_011511544.1 | 4 | 1020 | |||
XM_011513243.2 | XP_011511545.1 | 4 | 1020 | |||
XM_017007350.1 | XP_016862839.1 | 4 | 1020 | |||
XM_017007351.1 | XP_016862840.1 | 4 | 1020 | |||
XM_017007352.1 | XP_016862841.1 | 4 | 1020 | |||
GenBank mRNA | AK293745.1 | - | 4 | 638 | ||
AK303521.1 | - | 4 | 611 | |||
BC007504.2 | - | 4 | 601 | 2960711 | ||
DB454969.1 | - | 2 | 377 |