Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032782.4 | NP_116171.3 | 2 | 428 | ||
GenBank mRNA | AB924452.1 | - | 1 | 111 | ||
AF251707.1 | - | 2 | 300 | |||
AF450242.1 | - | 2 | 231 | |||
AF450243.1 | - | 2 | 231 | |||
AK027334.1 | - | 2 | 217 | |||
AK314406.1 | - | 2 | 235 | |||
AY069944.1 | - | 2 | 174 | |||
BC020843.1 | - | 2 | 283 | |||
BC063431.1 | - | 2 | 318 | 5229539 | ||
BP291914.1 | - | 2 | 428 | |||
JX049979.1 | - | 2 | 174 |