Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006166.3 | NP_006157.1 | 4 | 317 | ||
XM_005268908.4 | XP_005268965.1 | 5 | 381 | |||
XM_005268909.4 | XP_005268966.1 | 5 | 364 | |||
XM_005268910.4 | XP_005268967.1 | 5 | 1036 | |||
GenBank mRNA | AK291934.1 | - | 4 | 249 | ||
BC005316.1 | - | 4 | 266 | 3680937 | ||
BC005317.1 | - | 4 | 272 | 3997299 | ||
BC007035.1 | - | 4 | 244 | 3996244 | ||
BG776259.1 | - | 4 | 318 | |||
L06145.1 | - | 2 | 116 | |||
X59710.1 | - | 4 | 221 |