Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFATC2IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032815.3 | NP_116204.3 | 7 | 1082 | ||
GenBank mRNA | AF458593.1 | - | 7 | 1092 | ||
AK027545.1 | - | 4 | 497 | |||
AK074761.1 | - | 7 | 1072 | |||
AK297333.1 | - | 2 | 427 | |||
AK304606.1 | - | 2 | 349 | |||
AK316535.1 | - | 2 | 488 | |||
BC018311.1 | - | 5 | 586 | |||
BC021551.1 | - | 4 | 465 | |||
BC068007.1 | - | 4 | 322 | |||
BC080628.1 | - | 7 | 749 | |||
BC101741.1 | - | 7 | 1142 | 8069247 | ||
BC112182.1 | - | 7 | 1142 | 8327650 |