Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032664.3 | NP_116053.3 | 5 | 1909 | ||
XM_011544680.2 | XP_011542982.1 | 6 | 2375 | |||
GenBank mRNA | AJ431184.1 | - | 4 | 1721 | ||
AJ582015.1 | - | 5 | 1909 | |||
AK056199.1 | - | 2 | 395 | |||
AK292993.1 | - | 5 | 1907 | |||
AK297132.1 | - | 6 | 2002 | |||
AK303231.1 | - | 5 | 1842 | |||
BC063462.1 | - | 5 | 1949 | 5198060 |