Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRACR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144958.1 | NP_001138430.1 | 7, 8 | 1251 | ||
NM_032680.3 | NP_116069.1 | 7, 8 | 1251 | |||
XM_006719021.3 | XP_006719084.1 | 7, 8 | 1251 | |||
XM_011521034.2 | XP_011519336.1 | 6, 7 | 998 | |||
XM_011521036.2 | XP_011519338.1 | 5, 6 | 758 | |||
GenBank mRNA | AK091124.1 | - | 1, 2 | 87 | ||
AK304017.1 | - | 7, 8 | 1153 | |||
BC004524.2 | - | 7, 8 | 1148 | |||
BC150643.1 | - | 6, 7 | 882 | 9057013 |