Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032756.2 | NP_116145.1 | 1 | 1576 | ||
GenBank mRNA | BC007293.1 | - | 1 | 1544 | 3349473 |