Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316978.1 | NP_001303907.1 | 3 | 2518 | ||
NM_001316979.1 | NP_001303908.1 | 3 | 2245 | |||
NM_001316980.1 | NP_001303909.1 | 3 | 2068 | |||
NM_001316981.1 | NP_001303910.1 | 3 | 2370 | |||
NM_001316982.1 | NP_001303911.1 | 3 | 2188 | |||
NM_032792.3 | NP_116181.1 | 3 | 2274 | |||
XM_006723445.3 | XP_006723508.1 | 3 | 2288 | |||
GenBank mRNA | AK027392.1 | - | 3 | 2057 | ||
AK130056.1 | - | 4 | 1388 | |||
AL831909.1 | - | 2 | 819 | |||
BC024738.1 | - | 3 | 2269 | 5110354 |