Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032802.3 | NP_116191.2 | 6 | 814 | ||
XM_005254722.3 | XP_005254779.1 | 6 | 813 | |||
XM_017022680.1 | XP_016878169.1 | 6 | 813 | |||
XM_017022681.1 | XP_016878170.1 | 6 | 813 | |||
GenBank mRNA | AJ345028.1 | - | 6 | 792 | ||
AJ420896.1 | - | 4 | 365 | |||
AK027446.1 | - | 4 | 366 | |||
AK075454.1 | - | 6 | 803 | |||
AK315651.1 | - | 6 | 781 | |||
AY169314.1 | - | 6 | 810 | |||
BC025740.1 | - | 6 | 777 | 5223715 |