Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMGNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032806.5 | NP_116195.2 | 2 | 2363 | ||
XM_005265515.2 | XP_005265572.1 | 3 | 2494 | |||
XM_011534163.2 | XP_011532465.1 | 3 | 2482 | |||
XM_011534164.1 | XP_011532466.1 | 2 | 3064 | |||
XM_017007353.1 | XP_016862842.1 | 4 | 2573 | |||
GenBank mRNA | AK027472.1 | - | 1 | 1623 | ||
AK092147.1 | - | 2 | 2151 | |||
AK124737.1 | - | 1 | 3331 | |||
BC060861.1 | - | 2 | 2353 | 30335722 |