Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321967.1 | NP_001308896.1 | 10 | 1127 | ||
NM_001321968.1 | NP_001308897.1 | 9 | 1037 | |||
NM_001321969.1 | NP_001308898.1 | 11 | 1217 | |||
NM_032810.3 | NP_116199.2 | 10 | 1499 | |||
NR_135914.1 | 11 | 1214 | ||||
XM_005270252.4 | XP_005270309.1 | 10 | 1221 | |||
XM_011540302.1 | XP_011538604.1 | 8 | 892 | |||
XM_017016847.1 | XP_016872336.1 | 10 | 1368 | |||
XM_017016848.1 | XP_016872337.1 | 9 | 1371 | |||
XM_017016849.1 | XP_016872338.1 | 11 | 1340 | |||
XM_017016850.1 | XP_016872339.1 | 11 | 1391 | |||
GenBank mRNA | AF361493.1 | - | 9 | 1333 | ||
AK027506.1 | - | 7 | 727 | |||
AK075223.1 | - | 10 | 1428 | |||
AK304297.1 | - | 7 | 800 | |||
AL834370.1 | - | 9 | 912 | |||
BC010868.1 | - | 6 | 562 | |||
BC063530.1 | - | 9 | 958 | |||
BC073998.1 | - | 10 | 1157 | 6199813 |