Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032811.2 | NP_116200.2 | 9 | 2313 | ||
NR_016021.1 | 8 | 2176 | ||||
XM_005271709.3 | XP_005271766.1 | 6 | 2119 | |||
XM_017018442.1 | XP_016873931.1 | 6 | 3134 | |||
GenBank mRNA | AK027527.1 | - | 8 | 2147 | ||
AK027766.1 | - | 1 | 479 | |||
AK074140.1 | - | 8 | 1900 | |||
AL713631.1 | - | 5 | 1418 | |||
AL831881.1 | - | 3 | 1153 | |||
BC109269.2 | - | 7 | 1795 |