Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282933.1 | NP_001269862.1 | 13 | 2172 | ||
NM_001282935.1 | NP_001269864.1 | 12 | 1975 | |||
NM_032819.4 | NP_116208.3 | 13 | 2151 | |||
NR_104259.1 | 12 | 2075 | ||||
GenBank mRNA | AK027550.1 | - | 7 | 947 | ||
AK095848.1 | - | 1 | 1834 | |||
AK096755.1 | - | 11 | 1628 | |||
AK122943.1 | - | 13 | 2253 | |||
BC094738.1 | - | 12 | 1734 | |||
BC132873.1 | - | 13 | 1972 | 40146895 | ||
BC144188.1 | - | 13 | 1972 | 9052708 | ||
BC146811.1 | - | 13 | 1934 | 8860372 | ||
BC157823.1 | - | 13 | 1951 | 9057042 |