Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM87B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032824.2 | NP_116213.1 | 2 | 549 | ||
XM_005263827.2 | XP_005263884.1 | 2 | 506 | |||
XM_006712807.2 | XP_006712870.1 | 2 | 506 | |||
XM_017005121.1 | XP_016860610.1 | 2 | 52 | |||
GenBank mRNA | AK027587.1 | - | 2 | 534 | ||
AK290294.1 | - | 2 | 195 | |||
AY429534.1 | - | 2 | 52 | |||
AY429535.1 | - | 2 | 52 | |||
BC115373.1 | - | 2 | 353 | 40026807 | ||
DA806277.1 | - | 2 | 549 |