Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077268.1 | NP_001070736.1 | 7 | 1387 | ||
NM_001258421.1 | NP_001245350.1 | 7 | 1190 | |||
NM_032850.4 | NP_116239.3 | 8 | 1056 | |||
XM_017022683.1 | XP_016878172.1 | 6 | 1949 | |||
GenBank mRNA | AF445414.1 | - | 8 | 898 | ||
AK027746.1 | - | 7 | 819 | |||
AK122779.1 | - | 7 | 1177 | |||
AK225553.1 | - | 7 | 810 |