Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPND | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159846.2 | NP_001153318.1 | 3 | 407 | ||
NM_001300862.1 | NP_001287791.1 | 3 | 407 | |||
NM_032868.5 | NP_116257.2 | 3 | 407 | |||
XM_006722926.2 | XP_006722989.1 | 3 | 368 | |||
GenBank mRNA | AK027887.1 | - | 3 | 380 | ||
AK308468.1 | - | 3 | 368 | |||
BC032652.1 | - | 3 | 364 | 5503607 | ||
BP266054.1 | - | 3 | 367 | |||
BU150675.1 | - | 3 | 354 | |||
BX462788.2 | - | 3 | 407 |