Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PNISR SIRNA Assays ›
Gene Symbol:
PNISR
Gene Name:
PNN interacting serine and arginine rich protein
Gene Aliases:
C6orf111, HSPC306, SFRS18, SRrp130, bA98I9.2
Chromosome Location:
Chr.6: 99398051 - 99425345 on Build GRCh38
UniGene ID:
Hs.520287
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
34852
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PNISR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001322405.1
NP_001309334.1
3, 4
327
NM_001322406.1
NP_001309335.1
4, 5
443
NM_001322408.1
NP_001309337.1
2, 3
247
NM_001322410.1
NP_001309339.1
3, 4
331
NM_001322412.1
NP_001309341.1
3, 4
327
NM_001322413.1
NP_001309342.1
2, 3
251
NM_001322414.1
NP_001309343.1
3, 4
331
NM_001322415.1
NP_001309344.1
2, 3
251
NM_001322416.1
NP_001309345.1
3, 4
331
NM_001322417.1
NP_001309346.1
3, 4
331
NM_001322418.1
NP_001309347.1
2, 3
247
NM_001322419.1
NP_001309348.1
3, 4
331
NM_015491.2
NP_056306.1
2, 3
251
NM_032870.3
NP_116259.2
3, 4
331
NR_136326.1
2, 3
251
XM_005266912.3
XP_005266969.2
2, 3
210
XM_017010710.1
XP_016866199.1
3, 4
328
XM_017010711.1
XP_016866200.1
2, 3
214
XM_017010712.1
XP_016866201.1
3, 4
328
XM_017010713.1
XP_016866202.1
3, 4
324
GenBank mRNA
AF314184.1
-
1, 2
121
AF314186.1
-
3, 4
269
AJ420570.1
-
2, 3
1912
AK027658.1
-
3, 4
294
AK027759.1
-
3, 5
270
AK027898.1
-
3, 4
287
AK074628.1
-
2, 3
214
AK291155.1
-
1, 2
100
BC007791.2
-
3, 4
264
BC012780.2
-
3, 4
273
BC019074.2
-
3, 4
240
BC042850.1
-
4, 5
440
BC052638.1
-
3, 4
269
BC064413.1
-
3, 4
265
BC064640.1
-
3, 4
244
BC107700.1
-
3, 4
282
BX648520.1
-
2, 3
178
DA208838.1
-
3, 4
317
DA326969.1
-
3, 4
331
DB186090.1
-
4, 5
382
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-gx66z:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline