Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257133.1 | NP_001244062.1 | 3 | 560 | ||
NM_001257134.1 | NP_001244063.1 | 2 | 696 | |||
NM_032901.3 | NP_116290.1 | 2 | 500 | |||
GenBank mRNA | BC007849.2 | - | 2 | 364 | 4135996 | |
BG829783.1 | - | 2 | 430 | |||
BP369305.1 | - | 2 | 499 | |||
DW418935.1 | - | 3 | 353 |