Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC007854.2 | - | 10 | 1924 | 4298333 | |
BC049841.1 | - | 10 | 2176 | 4991947 | ||
BC053506.1 | - | 10 | 2265 | 2823232 |