Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190726.1 | NP_001177655.1 | 3, 4 | 540 | ||
NM_032918.2 | NP_116307.1 | 4, 5 | 597 | |||
XM_017020121.1 | XP_016875610.1 | 5, 6 | 1103 | |||
GenBank mRNA | AF339750.1 | - | 3, 4 | 340 | ||
AK295343.1 | - | 3, 4 | 480 | |||
AK313881.1 | - | 4, 5 | 537 | |||
BC007997.1 | - | 4, 5 | 490 | 3357341 |