Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006735.3 | NP_006726.1 | 2 | 898 | ||
GenBank mRNA | AK096742.1 | - | 1 | 1453 | ||
AL079274.1 | - | 1 | 239 | |||
BC130571.1 | - | 2 | 652 | 40146602 | ||
BC136500.1 | - | 2 | 652 | 9020489 | ||
BE465040.1 | - | 2 | 363 |