Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002929.2 | NP_002920.1 | 2, 3 | 943 | ||
XM_017020684.1 | XP_016876173.1 | 2, 3 | 2610 | |||
GenBank mRNA | AK289912.1 | - | 2, 3 | 1067 | ||
L77503.1 | - | 2, 3 | 850 | |||
U63973.1 | - | 2, 3 | 943 |