Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252678.1 | NP_001239607.1 | 2, 3 | 161 | ||
NM_001320665.1 | NP_001307594.1 | 3, 4 | 213 | |||
NM_033028.4 | NP_149017.2 | 3, 4 | 213 | |||
NR_045565.1 | 4, 5 | 271 | ||||
NR_045566.1 | 3, 4 | 526 | ||||
XM_011521848.1 | XP_011520150.1 | 3, 4 | 183 | |||
XM_011521849.1 | XP_011520151.1 | 3, 4 | 188 | |||
XM_011521851.1 | XP_011520153.1 | 3, 4 | 183 | |||
XM_017022450.1 | XP_016877939.1 | 3, 4 | 494 | |||
XM_017022451.1 | XP_016877940.1 | 2, 3 | 154 | |||
XM_017022452.1 | XP_016877941.1 | 3, 4 | 496 | |||
XM_017022453.1 | XP_016877942.1 | 3, 4 | 178 | |||
XM_017022454.1 | XP_016877943.1 | 2, 3 | 128 | |||
GenBank mRNA | AF359281.1 | - | 3, 4 | 185 | ||
AK075321.1 | - | 3, 4 | 177 | |||
AK298922.1 | - | 4, 5 | 243 | |||
AK302224.1 | - | 3, 4 | 480 | |||
AK303706.1 | - | 2, 3 | 156 | |||
AK314507.1 | - | 3, 4 | 185 | |||
AY457143.1 | - | 3, 4 | 285 | |||
BC027624.2 | - | 3, 4 | 183 | 4824276 | ||
BE727117.1 | - | 2, 3 | 154 | |||
BI562463.1 | - | 3, 4 | 203 | |||
BX647855.1 | - | 2, 3 | 132 | |||
DB269838.1 | - | 3, 4 | 206 | |||
DC320069.1 | - | 3, 4 | 208 | |||
DC372967.1 | - | 3, 4 | 213 | |||
KU177936.1 | - | 3, 4 | 164 |