Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136049.2 | NP_001129521.2 | 2 | 273 | ||
NM_033029.3 | NP_149018.2 | 2 | 273 | |||
NR_026786.1 | 2 | 152 | ||||
NR_026787.1 | 2 | 152 | ||||
XM_011513270.1 | XP_011511572.1 | 2 | 256 | |||
XM_011513271.1 | XP_011511573.1 | 2 | 256 | |||
XM_017007428.1 | XP_016862917.1 | 2 | 256 | |||
GenBank mRNA | AJ312398.1 | - | 2 | 152 | ||
AJ312399.1 | - | 2 | 273 | |||
AK299119.1 | - | 2 | 240 | |||
AL832783.1 | - | 2 | 258 | |||
AM920777.1 | - | 2 | 251 | |||
AM920778.1 | - | 2 | 95 |